missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-1-naphthylamine, 98%

Catalog No. p-183556
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147160250 25 g Glass Bottle
AC147162500 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147160250 Supplier Thermo Scientific Chemicals Supplier No. 147160250
Only null left

CAS: 90-30-2 | C16H13N | 219.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-30-2
Molecular Formula C16H13N
Molecular Weight (g/mol) 219.29
MDL Number MFCD00003878
InChI Key XQVWYOYUZDUNRW-UHFFFAOYSA-N
Synonym n-phenyl-1-naphthylamine, 1-anilinonaphthalene, pana, antioxidant pan, n-1-naphthyl aniline, neozone a, 1-naphthalenamine, n-phenyl, phenylnaphthylamine, nonox a, aceto pan
PubChem CID 7013
ChEBI CHEBI:34876
IUPAC Name N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color Brown to Pink
Boiling Point 226.0°C (15.0 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C10H7NHC6H5
Quantity 25 g
Beilstein 12, 1224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 219.29
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material N-Phenyl-1-naphthylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-983-0
RTECSNumber QM4500000
TSCA TSCA

RUO – Research Use Only