missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-1-anthramine 98.0+%, TCI America™

Catalog No. P1494500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1494500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1494500MG Supplier TCI America Supplier No. P1494500MG

Chemical Identifiers

CAS 98683-00-2
Molecular Formula C20H15N
Molecular Weight (g/mol) 269.347
InChI Key QCOIZKZASBFCJG-UHFFFAOYSA-N
Synonym 1-Anilinoanthracene
PubChem CID 21444085
IUPAC Name N-phenylanthracen-1-amine
SMILES C1=CC=C(C=C1)NC2=CC=CC3=CC4=CC=CC=C4C=C32

Specifications

Melting Point 162°C
Quantity 500 mg
Formula Weight 269.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenyl-1-anthramine