missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenacylpyridinium bromide, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 16883-69-5 |
|---|---|
| Molecular Formula | C13H12BrNO |
| Molecular Weight (g/mol) | 278.15 |
| InChI Key | PXSUMUYPXZEXDT-UHFFFAOYSA-M |
| Synonym | 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide |
| PubChem CID | 2798954 |
| IUPAC Name | 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide |
| SMILES | C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC450120100
|
N/A | 10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC450120500
|
N/A | 50g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 16883-69-5 | |
| 278.15 | |
| 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide | |
| 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide |
| C13H12BrNO | |
| PXSUMUYPXZEXDT-UHFFFAOYSA-M | |
| 2798954 | |
| C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-] |
Specifications
| 16883-69-5 | |
| 96% min. (Argentometry) | |
| 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide | |
| PXSUMUYPXZEXDT-UHFFFAOYSA-M | |
| 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide | |
| 2798954 | |
| 97% |
| Authentic | |
| C13H12BrNO | |
| Solubility in water: soluble. | |
| C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-] | |
| 278.15 | |
| 278.15 | |
| N-Phenacylpyridinium bromide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
GHS Signal Word: Warning
EINECSNumber : 240-919-6
RUO – Research Use Only