missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecanoic Acid, 98%, Spectrum™ Chemical

Chemical Identifiers

CAS 1002-84-2
Molecular Weight (g/mol) 242.4 g/mol
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612037
View Documents Promotion Details
N/A 5 g Poly Bottle N/A
Only null left
 
18612036
View Documents Promotion Details
N/A 25 g Poly Bottle N/A
Only null left
 
18612038
View Documents Promotion Details
N/A 500 g Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1002-84-2
242.4 g/mol

Specifications

1002-84-2
339°C
0.98
Crystalline Powder or Lumps
1
242.4 g/mol
Reagent
N-Pentadecanoic Acid
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.