missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecanoic Acid, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P235525GM

Quantity 25 g
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18612036


Only null left

Description

Description

Specifications

Chemical Identifiers

1002-84-2
242.4 g/mol

Specifications

N-Pentadecanoic Acid
1
Poly Bottle
242.4 g/mol
Reagent
1002-84-2
339°C
25 g
0.98
Crystalline Powder or Lumps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions