missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecanoic Acid, 98%, Spectrum™ Chemical

Catalog No. 18612038
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612038 500 g Poly Bottle
18612037 5 g Poly Bottle
18612036 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612038 Supplier Spectrum Chemical Mfg Cor Supplier No. P2355500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002-84-2
Molecular Weight (g/mol) 242.4 g/mol

Specifications

CAS Max % 1
Boiling Point 339°C
Packaging Poly Bottle
Quantity 500 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material N-Pentadecanoic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.