missing translation for 'onlineSavingsMsg'
Learn More

n-Paraffin Mix C18, C20, C22 & C24, Certified Reference Material, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022541
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022541 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022541 Supplier Merck Emd Millipore Supplier No. 47108

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
Flash Point Not Applicable
UN Number UN1262 - Class 3 - PG 2 - Octanes
Quantity 5 mL
Grade TraceCERT™ Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials