missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)-3-pyrroline 98.0+%, TCI America™

Catalog No. T16651G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16651G Supplier TCI America Supplier No. T16651G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16851-72-2
Molecular Formula C11H13NO2S
Molecular Weight (g/mol) 223.29
MDL Number MFCD01321189
InChI Key UNYMIBRUQCUASP-UHFFFAOYSA-N
Synonym N-Tosyl-3-pyrroline
PubChem CID 11138747
IUPAC Name 1-(4-methylphenyl)sulfonyl-2,5-dihydropyrrole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2CC=CC2

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 223.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-3-pyrroline