missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrichlorosilane 98.0+%, TCI America™

Catalog No. O016825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O016825G Supplier TCI America Supplier No. O016825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5283-66-9
Molecular Formula C8H17Cl3Si
Molecular Weight (g/mol) 247.659
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 229°C
Quantity 25 g
UN Number 1801
Formula Weight 247.66
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octyltrichlorosilane
EINECSNumber (2)-2041
RTECSNumber VV4700000
TSCA Yes