missing translation for 'onlineSavingsMsg'
Learn More

n-Octylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. O04521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04521G Supplier TCI America Supplier No. O04521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28741-08-4
Molecular Formula C8H19BO2
Molecular Weight (g/mol) 158.048
MDL Number MFCD01074560
PubChem CID 5195177
IUPAC Name octylboronic acid
SMILES B(CCCCCCCC)(O)O

Specifications

Melting Point 81°C
Color White
Quantity 1 g
Synonym n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid
InChI Key GKFRVXOKPXCXAK-UHFFFAOYSA-N
Formula Weight 158.05
Physical Form Crystalline Powder
Chemical Name or Material n-Octylboronic Acid (contains varying amounts of Anhydride)