missing translation for 'onlineSavingsMsg'
Learn More

n-Octylamine Hydrobromide 98.0+%, TCI America™

Catalog No. O04425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04425G 5 g
O04421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04425G Supplier TCI America Supplier No. O04425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14846-47-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
MDL Number MFCD00044802
InChI Key GIDDQKKGAYONOU-UHFFFAOYSA-N
Synonym n-Octylammonium Bromide
PubChem CID 53422655
IUPAC Name octan-1-amine;hydrobromide
SMILES CCCCCCCCN.Br

Specifications

Melting Point 208°C
Color White
Quantity 5 g
Formula Weight 210.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octylamine Hydrobromide