missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Isothiocyanate 98.0+%, TCI America™

Catalog No. O04645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04645G Supplier TCI America Supplier No. O04645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4430-45-9
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.30
MDL Number MFCD00041139
InChI Key YEZHGQZHWKJPCM-UHFFFAOYSA-N
Synonym octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
PubChem CID 78161
IUPAC Name 1-isothiocyanatooctane
SMILES CCCCCCCCN=C=S

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
UN Number 2810
Formula Weight 171.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Isothiocyanate