missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl beta-D-Glucopyranoside 96.0+%, TCI America™
$142.39 - $474.66
Chemical Identifiers
| CAS | 29836-26-8 |
|---|---|
| Molecular Formula | C14H28O6 |
| Molecular Weight (g/mol) | 292.37 |
| MDL Number | MFCD00063288 |
| InChI Key | HEGSGKPQLMEBJL-RKQHYHRCSA-N |
| Synonym | octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside |
| PubChem CID | 62852 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol |
| SMILES | CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
Chemical Identifiers
| 29836-26-8 | |
| 292.37 | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| 62852 | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| C14H28O6 | |
| MFCD00063288 | |
| octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol |
Specifications
| 29836-26-8 | |
| C14H28O6 | |
| octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O | |
| 292.37 | |
| 292.37 | |
| Crystalline Powder |
| White | |
| MFCD00063288 | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol | |
| 62852 | |
| ≥96.0% (GC) | |
| n-Octyl beta-D-Glucopyranoside |
Safety and Handling
EINECSNumber : (5)-6337&(5)-3641
TSCA : No