missing translation for 'onlineSavingsMsg'
Learn More

N-(o-Toluoyl)glycine 98,0+%, TCI America™

Numéro de catalogue. p-7136406
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T07195G 5 g
T071925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T07195G Fournisseur TCI America Code fournisseur T07195G

Identifiants chimiques

CAS 42013-20-7
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00050991
InChI Key YOEBAVRJHRCKRE-UHFFFAOYSA-N
Synonym 2-Methylhippuric Acid, N-(2-Methylbenzoyl)glycine
PubChem CID 91637
ChEBI CHEBI:68455
IUPAC Name Acide acétique 2-[(2-méthylbenzoyle)amino]acétique
SMILES CC1=CC=CC=C1C(=O)NCC(=O)O

Spécifications

Melting Point 163°C
Color Blanc
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material N-(o-Toluoyl)glycine