missing translation for 'onlineSavingsMsg'
Learn More

n-Nonane, 99%

Catalog No. AAA16177AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16177AP 500 mL
AAA16177AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16177AP Supplier Thermo Scientific Chemicals Supplier No. A16177AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-84-2 | C9H20 | 128.259 g/mol

Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

Solubility
Miscible with ethanol, ether, acetone, benzene, chloroform and hydrogen peroxide. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -53°C
Density 0.718
Boiling Point 150°C to 152°C
Flash Point 31°C (87°F)
Refractive Index 1.4054
Linear Formula CH3(CH2)7CH3
Quantity 500 mL
UN Number UN1920
Beilstein 1696917
Solubility Information Miscible with ethanol,ether,acetone,benzene,chloroform and hydrogen peroxide. Immiscible with water.
Formula Weight 128.26
Percent Purity 99%
Chemical Name or Material n-Nonane
Show More Show Less
Hazard Category H226-H304-H315-H319-H335-H336
Hazard Statement GHS H Statement
H304-H226-H332
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: NONANES
EINECSNumber 203-913-4
RTECSNumber RA6115000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only