missing translation for 'onlineSavingsMsg'
Learn More

n-Nonane, 99%, anhydrous, AcroSeal™

Catalog No. AC435631000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435631000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435631000 Supplier Thermo Scientific Chemicals Supplier No. 435631000

CAS: 111-84-2 | C9H20 | 128.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -53.0°C
Color Colorless
Density 0.7180g/mL
Boiling Point 151.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4040 to 1.4060
Linear Formula CH3(CH2)7CH3
Quantity 100 mL
Residue after Evaporation 0.0003% max.
Specific Gravity 0.718
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform
Formula Weight 128.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Nonane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 203-913-4

RUO – Research Use Only