missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraphenylbenzidine, 97%

Catalog No. AAH5965006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5965006 5 g
AAH5965003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5965006 Supplier Thermo Scientific Chemicals Supplier No. H5965006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 15546-43-7 | C36H28N2 | 488.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15546-43-7
Molecular Formula C36H28N2
Molecular Weight (g/mol) 488.63
MDL Number MFCD00228123
InChI Key DCZNSJVFOQPSRV-UHFFFAOYSA-N
Synonym n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
PubChem CID 84981
IUPAC Name N,N-diphenyl-4-[4-(N-phenylanilino)phenyl]aniline
SMILES C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 230°C to 234°C
Quantity 5 g
Formula Weight 488.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N,N,N',N'-Tetraphenylbenzidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-599-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only