missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. T29295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29295G 5 g
T292925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29295G Supplier TCI America Supplier No. T29295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 265651-18-1
Molecular Formula C9H16F6N3O3P
Molecular Weight (g/mol) 359.21
MDL Number MFCD01863753
InChI Key STWZCCVNXFLDDD-UHFFFAOYSA-N
Synonym hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
PubChem CID 16211151
IUPAC Name [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

Specifications

Melting Point 230°C
Color White
Quantity 5 g
Formula Weight 359.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium Hexafluorophosphate
TSCA No
Recommended Storage Freezer