missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals

Catalog No. AAJ64206MC
Change view
Click to view available options
Quantity:
100 mg
300 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64206MC 100 mg
AAJ64206EXH 300 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64206MC Supplier Thermo Scientific Chemicals Supplier No. J64206MC

A cell permeable and extremely high-affinity chelator of heavy metals

A cell permeable and extremely high-affinity chelator of heavy metals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16858-02-9
Molecular Formula C26H28N6
Molecular Weight (g/mol) 424.55
MDL Number MFCD00036918
InChI Key CVRXLMUYFMERMJ-UHFFFAOYSA-N
Synonym tpen, n,n,n',n'-tetrakis 2-pyridylmethyl ethylenediamine, n1,n1,n2,n2-tetrakis pyridin-2-ylmethyl ethane-1,2-diamine, unii-r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridylmethyl ethane-1,2-diamine, r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridyl-methyl ethylenediamine, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl-, oc-6-11, 1,2-ethanediamine,n,n,n',n'-tetrakis 2-pyridinylmethyl, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl
PubChem CID 5519
SMILES C(CN(CC1=CC=CC=N1)CC1=CC=CC=N1)N(CC1=CC=CC=N1)CC1=CC=CC=N1

Specifications

Melting Point 110°C to 112°C
Color White
Quantity 100 mg
Beilstein 4211982
IUPAC Name (2-{bis[(pyridin-2-yl)methyl]amino}ethyl)bis[(pyridin-2-yl)methyl]amine
Formula Weight 424.54
Physical Form Powder
Chemical Name or Material N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only