missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 50.0+%, TCI America™

Catalog No. T012025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T012025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T012025G Supplier TCI America Supplier No. T012025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-07-8
Molecular Formula C10H24N2O4
Molecular Weight (g/mol) 236.31
MDL Number MFCD00002854
InChI Key BYACHAOCSIPLCM-UHFFFAOYSA-N
Synonym theed, n,n,n',n'-tetrakis 2-hydroxyethyl ethylenediamine, ethylenediamine tetraethanol, entol, tked, ethylenedinitrilotetraethanol, tetrakis hydroxyethyl ethylenediamine, tetra hydroxyethyl ethylenediamine, unii-k5ape098zi, tetrahydroxyethyl ethylenediamine
PubChem CID 67322
IUPAC Name 2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol
SMILES OCCN(CCO)CCN(CCO)CCO

Specifications

Color Yellow
Quantity 25 g
Formula Weight 236.31
Percent Purity ≥50.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine
EINECSNumber (2)-1661
TSCA Yes