missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide 98.0+%, TCI America™

Catalog No. T28595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28595G 5 g
T285925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28595G Supplier TCI America Supplier No. T28595G

Chemical Identifiers

CAS 6334-25-4
Molecular Formula C14H28N2O6
Molecular Weight (g/mol) 320.39
MDL Number MFCD07787618
InChI Key OKRNLSUTBJUVKA-UHFFFAOYSA-N
PubChem CID 95283
IUPAC Name N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide
SMILES OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 5 g
Synonym N,N,N′C,N′C-Tetrakis(2-hydroxyethyl)hexanediamide
Formula Weight 320.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(2-hydroxyethyl)adipamide
EINECSNumber (2)-3798
TSCA No