missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic Acid) 98.0+%, TCI America™

Catalog No. E0393500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0393500G 500 g
E039325G 25 g
E0393100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0393500G Supplier TCI America Supplier No. E0393500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1429-50-1
Molecular Formula C6H20N2O12P4
Molecular Weight (g/mol) 436.123
MDL Number MFCD00058893
InChI Key NFDRPXJGHKJRLJ-UHFFFAOYSA-N
Synonym edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid
PubChem CID 15025
IUPAC Name [2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid
SMILES C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O

Specifications

Color White
pH <2 (1% H2O soln. 25°C)
Quantity 500 g
Formula Weight 436.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Ethylenediaminetetrakis(methylenephosphonic Acid)
EINECSNumber (2)-1873
TSCA Yes