missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99%

Catalog No. AC204661000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204661000 100 g Glass bottle
AC204660050 5 g Glass bottle
AC204660250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204661000 Supplier Thermo Scientific Chemicals Supplier No. 204661000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2491-06-7 | C4H9NO2·HCl | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2491-06-7
Molecular Formula C4H9NO2·HCl
Molecular Weight (g/mol) 139.58
MDL Number MFCD00012610
InChI Key FKASAVXZZLJTNX-UHFFFAOYSA-N
Synonym n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride
PubChem CID 75605
IUPAC Name 2-(dimethylamino)acetic acid;hydrochloride
SMILES CN(C)CC(=O)O.Cl

Specifications

Melting Point 188°C to 191°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CO2H·HCl
Beilstein 04, I, 469
Merck Index 15, 3269
Solubility Information Solubility in water: 100g/L (20 c). Other solubilities: insoluble in chloroform and acetone
Formula Weight 139.58
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material N, N-Dimethylglycine hydrochloride
Show More Show Less
EINECSNumber 219-648-2

RUO – Research Use Only