missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Dipropyl Acetal 90.0+%, TCI America™

Catalog No. D13015ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13015ML 5 mL
D130125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13015ML Supplier TCI America Supplier No. D13015ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6006-65-1
Molecular Formula C9H22NO2
Molecular Weight (g/mol) 176.28
MDL Number MFCD00009374
InChI Key NSLGQFIDCADTAS-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dipropyl acetal, 1,1-dipropoxytrimethylamine, n,n-dimethylformamide di-n-propyl acetal, dipropoxymethyl dimethylamine, 1,1-dipropoxy-n,n-dimethylmethylamine, acmc-1b5t3, dimethyl dipropoxymethyl amine, dimethylformamide dipropyl acetal, nslgqfidcadtas-uhfffaoysa, n,n-dimethylformamide dipropylacetal
PubChem CID 80105
IUPAC Name (dipropoxymethyl)dimethylazanium
SMILES CCCOC(OCCC)[NH+](C)C

Specifications

Color Yellow
Quantity 5 mL
UN Number 1993
Formula Weight 175.27
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dipropyl Acetal [for Esterification]