missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide diethyl acetal, 95%

Catalog No. AC162080500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162080500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162080500 Supplier Thermo Scientific Chemicals Supplier No. 162080500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1188-33-6 | C7H17NO2 | 147.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 1188-33-6
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
MDL Number MFCD00009227
InChI Key BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
PubChem CID 70913
IUPAC Name 1,1-diethoxy-N,N-dimethylmethanamine
SMILES CCOC(N(C)C)OCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.8590g/mL
Boiling Point 128°C to 130°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH(OC2H5)2
Quantity 50 g
Fieser 01,281; 02,154; 05,253; 07,125; 10,157
Specific Gravity 0.859
Solubility Information Solubility in water: decomposes
Formula Weight 147.22
Percent Purity 95%
Chemical Name or Material N,N-Dimethylformamide diethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 214-707-9

RUO – Research Use Only