missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecylamine 95.0+%, TCI America™

Catalog No. D2288500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2288500ML 500 mL
D228825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2288500ML Supplier TCI America Supplier No. D2288500ML

Chemical Identifiers

CAS 1120-24-7
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00015060
InChI Key YWWNNLPSZSEZNZ-UHFFFAOYSA-N
Synonym 1-(Dimethylamino)decane
PubChem CID 70711
IUPAC Name decyldimethylamine
SMILES CCCCCCCCCCN(C)C

Specifications

Color Colorless
Boiling Point 235°C
Quantity 500 mL
UN Number 2735
Formula Weight 185.36
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyldecylamine
EINECSNumber (2)-0176
TSCA Yes