missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecanamide, 98%

Catalog No. AAL0873114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0873114 25 g
AAL0873106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0873114 Supplier Thermo Scientific Chemicals Supplier No. L0873114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14433-76-2 | C12H25NO | 199.338 g/mol

N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 14433-76-2
Molecular Formula C12H25NO
Molecular Weight (g/mol) 199.338
MDL Number MFCD00043725
InChI Key HNXNKTMIVROLTK-UHFFFAOYSA-N
Synonym decanamide, n,n-dimethyl, n,n-dimethylcapramide, n,n-dimethylcapylamide, n,n-dimethyldecanoamide, unii-o29y6x2jez, n,n-dimethyldecan-1-amide, nn-dimethyldecanamide, n,n-dimethyl decanamide, o29y6x2jez, dimethyl capramide
PubChem CID 26690
IUPAC Name N,N-dimethyldecanamide
SMILES CCCCCCCCCC(=O)N(C)C

Specifications

Boiling Point 110°C to 111°C (0.5 mmHg)
Refractive Index 1.454
Quantity 25 g
Beilstein 1906042
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 199.34
Percent Purity 98%
Chemical Name or Material N,N-Dimethyldecanamide
EINECSNumber 238-405-1
RTECSNumber HD6350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only