missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized

Catalog No. p-3778342
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264130250 25 g Glass bottle
AC264131000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264130250 Supplier Thermo Scientific Chemicals Supplier No. 264130250

CAS: 18871-66-4 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18871-66-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008476
InChI Key FBZVZUSVGKOWHG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332
PubChem CID 87835
IUPAC Name 1,1-dimethoxy-N,N-dimethylethanamine
SMILES CC(N(C)C)(OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9110g/mL
Boiling Point 110.0°C to 118.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Quantity 25 g
Fieser 04,166; 05,226; 10,154
Specific Gravity 0.911
Stabilizer sodium carbonate
Formula Weight 133.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacetamide dimethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and eas
EINECSNumber 242-641-

RUO – Research Use Only