missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020380
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020380 1 mL
111018782 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020380 Supplier Merck Emd Millipore Supplier No. 723361MLF

N,N-Dimethylacetamide is a highly polar solvent, which can find applications in a variety of chemical reactions. A mixture of lithium chloride in N,N-dimethylacetamide may be used for the dissolution of cellulose to form a homogeneous solution.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 914°F
CAS 127-19-5
Vapor Pressure 2 mmHg (25°C)
Vapor Density 3 (vs. air)
Density 0.937 g/mL (at 25°C)
Flash Point 64°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CON(CH3)2
MDL Number MFCD00008686
Melting Point -20°C
Boiling Point 164.5°C to 166°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 87.12
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less