missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine, 97%

Catalog No. AC408465000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408465000 500 g Glass bottle
AC408460050 5 g Glass bottle
AC408460250 25 g Glass bottle
AC408461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC408465000 Supplier Thermo Scientific Chemicals Supplier No. 408465000

CAS: 99-98-9 | C8H12N2 | 136.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-98-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007860
InChI Key BZORFPDSXLZWJF-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline
PubChem CID 7472
ChEBI CHEBI:15783
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(N)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35.0°C
Color Brown
Density 1.0900g/mL
Boiling Point 262.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2C6H4N(CH3)2
Quantity 500 g
Specific Gravity 1.09
Merck Index 15, 3278
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: soluble in alcohol,chloroform,ether,benzene
Formula Weight 136.2
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine, Practical
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
EINECSNumber 202-807-5

RUO – Research Use Only