missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-m-anisidine 97.0+%, TCI America™
Supplier: TCI America D128825ML
Specifications
| N,N-Dimethyl-m-anisidine | |
| Yellow | |
| C9H13NO | |
| 25 mL | |
| MOYHVSKDHLMMPS-UHFFFAOYSA-N | |
| 3-methoxy-N,N-dimethylaniline | |
| 139977 | |
| ≥97.0% (GC,T) |
| 15799-79-8 | |
| 123°C | |
| MFCD00051779 | |
| 3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine | |
| CN(C)C1=CC(=CC=C1)OC | |
| 151.209 | |
| 151.21 | |
| Liquid |
Chemical Identifiers
| 15799-79-8 | |
| 151.209 | |
| MOYHVSKDHLMMPS-UHFFFAOYSA-N | |
| 139977 | |
| CN(C)C1=CC(=CC=C1)OC |
| C9H13NO | |
| MFCD00051779 | |
| 3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine | |
| 3-methoxy-N,N-dimethylaniline |
Safety and Handling
TSCA : No