accessibility menu, dialog, popup

UserName

CH3ONCH3H3C

CAS RN 15799-79-8

IUPAC Name: 3-methoxy-N,N-dimethylaniline
Synonyms: 3-methoxy-N,N-dimethylaniline
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: MOYHVSKDHLMMPS-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)N(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)