missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitroaniline, 98+%

Catalog No. AAL0040406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0040406 5 g
AAL0040414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0040406 Supplier Thermo Scientific Chemicals Supplier No. L0040406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-23-2 | C8H10N2O2 | 166.18 g/mol

It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.

Solubility
Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 100-23-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014712
InChI Key QJAIOCKFIORVFU-UHFFFAOYSA-N
Synonym 4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro
PubChem CID 7491
IUPAC Name N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 163°C to 166°C
Quantity 5 g
UN Number UN2811
Beilstein 638087
Solubility Information Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethyl-4-nitroaniline
Hazard Category H301+H311+H331-H315-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 202-832-1
RTECSNumber BX7035000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only