missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-dodecylamine N-oxide, 95%

Catalog No. AAH2606506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606506 5 g
AAH2606514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2606506 Supplier Thermo Scientific Chemicals Supplier No. H2606506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1643-20-5 | C14H31NO | 229.408 g/mol

N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1643-20-5
Molecular Formula C14H31NO
Molecular Weight (g/mol) 229.408
MDL Number MFCD00002049
InChI Key SYELZBGXAIXKHU-UHFFFAOYSA-N
Synonym lauramine oxide, lauryldimethylamine oxide, dodecyldimethylamine oxide, lauryldimethylamine n-oxide, ldao, ammonyx ao, ammonyx lo, empigen ob, aromox dmcd, conco xal
PubChem CID 15433
IUPAC Name N,N-dimethyldodecan-1-amine oxide
SMILES CCCCCCCCCCCC[N+](C)(C)[O-]

Specifications

Melting Point 120°C to 123°C
Density 0.996
Quantity 5 g
UN Number UN3259
Beilstein 1769927
Sensitivity Very Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 229.4
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-1-dodecylamine N-oxide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 216-700-6
RTECSNumber JR6650000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only