missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,3-propanediamine, 99%

Catalog No. p-7043934
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03539AK 250 mL
AAL03539AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL03539AK Supplier Thermo Scientific Chemicals Supplier No. L03539AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-55-7 | C5H14N2 | 102.181 g/mol

Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 109-55-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00008216
InChI Key IUNMPGNGSSIWFP-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
PubChem CID 7993
IUPAC Name N',N'-dimethylpropane-1,3-diamine
SMILES CN(C)CCCN

Specifications

Melting Point -70°C
Density 0.818
Boiling Point 126°C to 128°C
Flash Point 32°C (89°F)
Odor Ammonia-like
Refractive Index 1.435
Linear Formula (CH3)2N(CH2)3NH2
Quantity 250 mL
UN Number UN2734
Beilstein 605293
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-1,3-propanediamine
Show More Show Less
Hazard Category H226-H302+H312-H314-H317-H335
Hazard Statement GHS H Statement
H314-H226-H302-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 203-680-9
RTECSNumber TX7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only