missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylenediamine, 97%

Catalog No. AAL0476509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0476509 10 g
AAL0476518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0476509 Supplier Thermo Scientific Chemicals Supplier No. L0476509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 121-05-1 | C8H20N2 | 144.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air, oxidizing agents. Incompatible materials are Strong oxidizing agents, Strong acids

Chemical Identifiers

CAS 121-05-1
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00015015
InChI Key CURJNMSGPBXOGK-UHFFFAOYSA-N
Synonym 2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
PubChem CID 8459
IUPAC Name N',N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)N(CCN)C(C)C

Specifications

Density 0.828
Boiling Point 170°C
Flash Point 54°C (129°F)
Refractive Index 1.442
Quantity 10 g
UN Number UN2733
Beilstein 1734800
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N,N-Diisopropylethylenediamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 204-447-4
RTECSNumber KV4200000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only