missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylaniline, 99%

Catalog No. AAA10560AP
Change view
Click to view available options
Quantity:
10,000 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10560AP 500 mL
AAA105600F 2500 mL
AAA105600D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10560AP Supplier Thermo Scientific Chemicals Supplier No. A10560AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-66-7 | C10H15N | 149.24 g/mol

N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.      

Notes
Incompatible with strong oxidizing agents and strong acids.                  .

Chemical Identifiers

CAS 91-66-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00009042,MFCD31699978
InChI Key GGSUCNLOZRCGPQ-UHFFFAOYSA-N
Synonym diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine
PubChem CID 7061
IUPAC Name N,N-diethylaniline
SMILES CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -38°C
Density 0.936
Boiling Point 215°C to 216°C
Flash Point 85°C (185°F)
Refractive Index 1.542
Quantity 500 mL
UN Number UN2432
Beilstein 742483
Merck Index 14,3114
Solubility Information Miscible with water and acetone. Slightly miscible with chloroform,alcohol and ether.
Formula Weight 149.24
Percent Purity 99%
Chemical Name or Material N,N-Diethylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N,N DIETHYLANILINE
EINECSNumber 202-088-8
RTECSNumber BX3400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only