missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™

Catalog No. D36015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36015G 5 g
D36011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36015G Supplier TCI America Supplier No. D36015G

Chemical Identifiers

CAS 920304-57-0
Molecular Formula C16H26BNO2
Molecular Weight (g/mol) 275.20
MDL Number MFCD10000956
InChI Key WVPMYKCIKQQJIV-UHFFFAOYSA-N
Synonym 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Diethylamino)phenylboronic Acid Pinacol Ester
PubChem CID 44629855
IUPAC Name N,N-diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CCN(CC)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Color White
Quantity 5 g
Formula Weight 275.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline