missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-2,3-dimethylbenzamide 98.0+%, TCI America™
Supplier: TCI America D295625G
Specifications
| N,N-Diethyl-2,3-dimethylbenzamide | |
| Yellow | |
| Clear Liquid at 20°C | |
| MFCD06797096 | |
| SMVYLWURFNYXTP-UHFFFAOYSA-N | |
| N,N-diethyl-2,3-dimethylbenzamide | |
| 15621069 | |
| ≥98.0% (GC) |
| 57806-76-5 | |
| 25 g | |
| C13H19NO | |
| N,N-Diethyl-o-xylene-3-carboxamide | |
| CCN(CC)C(=O)C1=C(C(=CC=C1)C)C | |
| 205.301 | |
| 205.30 |
Chemical Identifiers
| 57806-76-5 | |
| 205.301 | |
| SMVYLWURFNYXTP-UHFFFAOYSA-N | |
| 15621069 | |
| CCN(CC)C(=O)C1=C(C(=CC=C1)C)C |
| C13H19NO | |
| MFCD06797096 | |
| N,N-Diethyl-o-xylene-3-carboxamide | |
| N,N-diethyl-2,3-dimethylbenzamide |
Safety and Handling
TSCA : No