accessibility menu, dialog, popup

UserName

H3CNCH3OCH3CH3

CAS RN 57806-76-5

IUPAC Name: N,N-diethyl-2,3-dimethylbenzamide
Synonyms: N,N-diethyl-2,3-dimethylbenzamide
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: SMVYLWURFNYXTP-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=C(C)C(C)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results