missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2,3-dimethylbenzamide 98.0+%, TCI America™

Catalog No. p-7127582
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29565G 5 g
D295625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29565G Supplier TCI America Supplier No. D29565G

Chemical Identifiers

CAS 57806-76-5
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD06797096
InChI Key SMVYLWURFNYXTP-UHFFFAOYSA-N
Synonym N,N-Diethyl-o-xylene-3-carboxamide
PubChem CID 15621069
IUPAC Name N,N-diethyl-2,3-dimethylbenzamide
SMILES CCN(CC)C(=O)C1=C(C(=CC=C1)C)C

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Formula Weight 205.30
Percent Purity ≥98.0% (GC)
Chemical Name or Material N,N-Diethyl-2,3-dimethylbenzamide