missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2,3,3,3-tetrafluoropropionamide, 95%

Catalog No. AAL1673914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1673914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1673914 Supplier Thermo Scientific Chemicals Supplier No. L1673914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392-63-2 | C7H11F4NO | 201.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 392-63-2
Density 1.228
Boiling Point 58°C (5 mmHg)
Molecular Formula C7H11F4NO
Refractive Index 1.392
Linear Formula CF3CHFCON(CH2CH3)2
Quantity 25 g
MDL Number MFCD00069160
Beilstein 1780406
Synonym n,n-diethyl-2,3,3,3-tetrafluoropropionamide, acmc-20ak99, n,n-diethyl-2,3,3,3-tetrafluoropropanamid, n,n-diethyl-2,3,3,3-tetrafluoropropioamide, n,n-diethyl-2,2,3,3-tetrafluoro-propionamide, propanamide,n,n-diethyl-2,3,3,3-tetrafluoro, n,n-diethyl-2,3,3,3-tetrakis fluoranyl propanamide
InChI Key OIZFBXRWENZGCS-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)C(F)(F)C(F)F
IUPAC Name N,N-diethyl-2,2,3,3-tetrafluoropropanamide
Molecular Weight (g/mol) 201.17
PubChem CID 2774042
Formula Weight 201.16
Percent Purity 95%
Chemical Name or Material N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only