missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-cyclohexanediamine (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. D45665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45665G 5 g
D45661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45665G Supplier TCI America Supplier No. D45665G

Chemical Identifiers

CAS 42389-54-8
Molecular Formula C10H22N2
Molecular Weight (g/mol) 170.3
MDL Number MFCD06658428
InChI Key ZFWPGRUTHAZDBK-UHFFFAOYSA-N
Synonym 1-Amino-4-(diethylamino)cyclohexane
PubChem CID 349884
IUPAC Name 4-N,4-N-diethylcyclohexane-1,4-diamine
SMILES CCN(CC)C1CCC(CC1)N

Specifications

Color Colorless
Boiling Point 111°C
Quantity 5 g
UN Number 2735
Formula Weight 170.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,4-cyclohexanediamine (cis- and trans- mixture)