missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-1,3-diaminopropane 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 171°C |
| Quantity | 500 mL |
| UN Number | 2684 |
| Formula Weight | 130.24 |
| Percent Purity | ≥99.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Diethyl-1,3-diaminopropane |
Safety and Handling
| EINECSNumber | (2)-0158 |
| RTECSNumber | TX7350000 |
| TSCA | Yes |