missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dicyclohexylmethylamine, 97%, Thermo Scientific™

Numéro de catalogue. AC428392500
Modifier l'affichage
Click to view available options
Quantité:
250g
1kg
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC428392500 250g Glass bottle
AC428390010 1kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC428392500 Fournisseur Thermo Scientific Chemicals Code fournisseur 428392500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 7560-83-0
Formule moléculaire C13H25N
Poids moléculaire (g/mol) 195.35
Numéro MDL MFCD00014289
Clé InChI GSCCALZHGUWNJW-UHFFFAOYSA-N
Synonyme n,n-dicyclohexylmethylamine, n-methyldicyclohexylamine, cyclohexanamine, n-cyclohexyl-n-methyl, dicyclohexylamine, n-methyl, n-methyl dicyclohexylamine, unii-ri95g6j8ff, n-cyclohexyl-n-methylcyclohexylamine, cy2nme, methyldicyclohexylamine, nn-dicyclohexylmethylamine
PubChem CID 24210
Nom de l’IUPAC N-cyclohexyl-N-methylcyclohexanamine
SOURIRES CN(C1CCCCC1)C2CCCCC2

Spécifications

Nom ou substance chimique N, N-Dicyclohexylmethylamine
CAS % minimum 96.0
CAS % maximal 100.0
Densité 0.9210g/mL
Point d’ébullition 265.0°C
Point d’éclair 110°C
Fourchette de pourcentages d’essai 96% min. (GC)
Emballage Glass bottle
Quantité 250g
Gravité spécifique 0.921
Poids de formule 195.35
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-453-4

RUO – Research Use Only