missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™

Catalog No. AC429021000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429021000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC429021000 Supplier Thermo Scientific Chemicals Supplier No. 429021000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-75-0 | C13H22N2 | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Specifications

Chemical Name or Material N, N'-Dicyclohexylcarbodiimide
CAS 538-75-0, 75-09-2
CAS Min % 15.81, 81.7
CAS Max % 18.3, 84.19
Color White to Brown
Density 1.2400g/mL
Flash Point 87°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Molecular Formula C13H22N2
Linear Formula C6H11N=C=NC6H11
Quantity 100 mL
MDL Number MFCD00011659
Specific Gravity 1.24
Merck Index 15, 3107
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILES C1CCC(CC1)N=C=NC2CCCCC2
IUPAC Name N,N'-dicyclohexylmethanediimine
Molecular Weight (g/mol) 206.33
PubChem CID 10868
ChEBI CHEBI:53090
Formula Weight 206.33
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only