missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibutylaniline, 97%

Catalog No. AAL1339006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1339006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1339006 Supplier Thermo Scientific Chemicals Supplier No. L1339006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 613-29-6 | C14H23N | 205.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-29-6
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00035799
InChI Key FZPXKEPZZOEPGX-UHFFFAOYSA-N
Synonym benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl
PubChem CID 61154
IUPAC Name N,N-dibutylaniline
SMILES CCCCN(CCCC)C1=CC=CC=C1

Specifications

Density 0.906
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Refractive Index 1.518
Quantity 5 g
UN Number UN2810
Beilstein 2210422
Formula Weight 205.35
Percent Purity 97%
Chemical Name or Material N,N-Dibutylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 210-335-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only