missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine Diacetate 97.0+%, TCI America™

Catalog No. D028025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D028025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D028025G Supplier TCI America Supplier No. D028025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-75-8
Molecular Formula C20H28N2O4
Molecular Weight (g/mol) 360.454
MDL Number MFCD00040588
InChI Key MTRNNCLQPVCDLF-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
PubChem CID 31228
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 360.45
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dibenzylethylenediamine Diacetate
RTECSNumber KV3400000
TSCA Yes