missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dibenzyl-4,13-diaza-18-crown 6-Ether 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 84°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | N,N′-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane |
| Formula Weight | 442.60 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Dibenzyl-4,13-diaza-18-crown 6-Ether |
Sécurité et manipulation
| TSCA | No |