missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)thiourea, 98%

CAS: 1516-32-1 | C5H12N2S | 132.23 g/mol

Supplier:  Thermo Scientific Chemicals L0965409

 View more versions of this product

Catalog No. AAL0965409

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(n-Butyl)thiourea
76°C to 79°C
C5H12N2S
10 g
1-butyl-2-thiourea, n-butylthiourea, thiourea, butyl, n-butyl thiourea, butyl-2-thiourea, thiourea, n-butyl, usaf d-5, 1-butylthiourea, urea, 1-butyl-2-thio, n-n-butyl thiourea
CCCCNC(N)=S
132.23
132.23
1516-32-1
Odorless
MFCD00022173
1744779
GMEGXJPUFRVCPX-UHFFFAOYSA-N
butylthiourea
1551919
98%

Chemical Identifiers

1516-32-1
132.23
GMEGXJPUFRVCPX-UHFFFAOYSA-N
1551919
CCCCNC(N)=S
C5H12N2S
MFCD00022173
1-butyl-2-thiourea, n-butylthiourea, thiourea, butyl, n-butyl thiourea, butyl-2-thiourea, thiourea, n-butyl, usaf d-5, 1-butylthiourea, urea, 1-butyl-2-thio, n-n-butyl thiourea
butylthiourea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 216-165-9

RTECSNumber : YS4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only